missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4A Antibody (741531), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4694PEATT594
This item is not returnable.
View return policy
Description
Semaphorin 4A Monoclonal antibody specifically detects Semaphorin 4A in Human samples. It is validated for Flow Cytometry
Specifications
| Semaphorin 4A | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 741531 | |
| Flow Cytometry | |
| CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287 | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4A, Gly32-His683, Accession # Q9H3S1 | |
| 0.1 mL | |
| Neuroscience, Vision | |
| 64218 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction