missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00
Specifications
| Antigen | Semaphorin 4A |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Semaphorin 4A Polyclonal specifically detects Semaphorin 4A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Semaphorin 4A | |
| Polyclonal | |
| Rabbit | |
| Neuroscience, Vision | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 64218 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PNLNSWKQDMERGNPEWACASGPMSRSLRPQSRPQIIKEVLAVPNSILELPCPHLSALASYYWSHGPAAVPEASSTVYNGSLLLIVQDGVGGLYQCWATENGFSYPVISYWVDSQDQTLALDPELAGIPREHVKVPLTRVSGGA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CORD10, RP35, Sema B, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4A, SEMAB, semaphorin-4A, Semaphorin-B, SemB, SEMBFLJ12287 | |
| SEMA4A | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title