missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4C Antibody (663203) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8497JF635
This item is not returnable.
View return policy
Description
Semaphorin 4C Monoclonal antibody specifically detects Semaphorin 4C in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Semaphorin 4C | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C, Ala21-Phe664, Accession # Q64151 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 663203 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Neuroscience | |
| 54910 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction