Learn More
Semaphorin 4C Antibody (663203), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB8497PECY7
Product Code. 30026051
  • € 640.00 / 0.10mL
Description
Semaphorin 4C Monoclonal antibody specifically detects Semaphorin 4C in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Semaphorin 4C
Monoclonal
PE-Cyanine7
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry
663203
Flow Cytometry
FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C, Ala21-Phe664, Accession # Q64151
0.1 mL
Neuroscience
54910
Store at 4C in the dark. Do not freeze.