missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (A8), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25198AF594
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Monoclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Flow (Cell Surface)
Specifications
| Semaphorin 4D/CD100 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody. | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 10507 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunoprecipitation, Flow Cytometry (Cell Surface) | |
| A8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur