missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93561APC
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Polyclonal antibody specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Western Blot
Specifications
| Semaphorin 4D/CD100 | |
| Polyclonal | |
| Western Blot | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| Synthetic peptide corresponding to residues 147-158 [GKNEDGKGRCPF] of the human CD100 protein. | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 10507 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto