missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4F Antibody (780225) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7200JF525
This item is not returnable.
View return policy
Description
Semaphorin 4F Monoclonal antibody specifically detects Semaphorin 4F in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
Specifications
| Semaphorin 4F | |
| Monoclonal | |
| Janelia Fluor 525 | |
| M-SEMA, m-Sema-M, PRO2353, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4F, Sema M, Sema W, SEMAM, semaphorin-4F, semaphorin-M, Semaphorin-W, SEMAWsemaphorin M | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F, Arg41-Gly667, Accession # Q9Z123 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 780225 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Neuroscience | |
| 10505 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction