missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 6B Antibody (MM0535-10F7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11880AF594
This item is not returnable.
View return policy
Description
Semaphorin 6B Monoclonal specifically detects Semaphorin 6B in Human samples. It is validated for Western Blot.
Specifications
| Semaphorin 6B | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SEMA6B | |
| Human recombinant Semaphorin 6B (extracellular domain) | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| MM0535-10F7 | |
| Western Blot | |
| sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B, SEMAN, semaphorin VIB, semaphorin Z, semaphorin-6B, semaphorin-6Ba, SEMA-VIB, semaZ, SEM-SEMA-Y, UNQ1907/PRO4353 | |
| Mouse | |
| 0.1 mL | |
| 10501 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction