Learn More
B Braun™ Semken Delicate Dissecting Forceps
Product Code. 12009830
Change view
Click to view available options
Length (Metric):
125 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12009830 125 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12009830 Supplier B Braun™ Supplier No. BD215R
  • ONLINE EXCLUSIVE PRICE
    € 26.05
    € 24.25 / Pack of 1
    Save € 1.80
    6% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Lightweight, narrow and toothed forceps for handling fine membranes or single layer tissues.
  • Serrated tip
  • Length of 125 mm
  • Manufactured from stainless steel
Specifications
Product Type Forceps
Material Stainless Steel
For Use With (Application) Surgery/Medical
Shape Serrated
Style Semken
Length (Metric) 125 mm