missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody (5D8), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52727AF700
This item is not returnable.
View return policy
Description
Senataxin Monoclonal antibody specifically detects Senataxin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular)
Specifications
| Senataxin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| Partial recombinant human Senataxin protein (between amino acids 2300-2600) [UniProt Q7Z333] | |
| 0.1 mL | |
| DNA Repair | |
| 23064 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| 5D8 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction