missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57542CL1
This item is not returnable.
View return policy
Description
Senataxin Polyclonal antibody specifically detects Senataxin in Human samples. It is validated for Immunoprecipitation,Western Blot,ChIP assay (ChIP),KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| Senataxin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, ChIP Assay, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human senataxin using the numbering given in entry NP_055861.2 (GeneID 23064). | |
| 0.1 mL | |
| DNA Repair | |
| 23064 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction