missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57543R
This item is not returnable.
View return policy
Description
Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for Western Blot, Knockout Validated, Knockdown Validated.
Specifications
| Senataxin | |
| Polyclonal | |
| Western Blot, Knockout Validated, KnockDown Validated | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoassay, KnockDown | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SETX | |
| The immunogen recognized by this antibody maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2). | |
| 100 μL | |
| DNA Repair | |
| 23064 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction