missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57543DL594
This item is not returnable.
View return policy
Description
Senataxin Polyclonal antibody specifically detects Senataxin in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative), ChIP assay (ChIP), KnockDown
Specifications
| Senataxin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative), ChIP Assay, KnockDown | |
| DyLight 594 | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| The immunogen recognized by this antibody maps to a region between residue 2627 and 2677 of human senataxin (NP_055861.2). | |
| 0.1 mL | |
| DNA Repair | |
| 23064 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction