missing translation for 'onlineSavingsMsg'
Learn More

Senicapoc, MedChemExpress

Product Code. 16438088
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16438088

Brand: Medchem Express HY50694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Senicapoc (ICA-17043) is a potent and selective Gardos channel (Ca2+-activated K+ channel; KCa3.1) blocker with an IC50 of 11 nM. Senicapoc blocks Ca2+-induced rubidium flux from human RBCs with an IC50 value of 11 nM and inhibits RBC dehydration with IC50 of 30 nM.

Senicapoc (ICA-17043) is a potent and selective Gardos channel (Ca2+-activated K+ channel

Specifications

Chemical Name or Material Senicapoc
CAS 289656-45-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H15F2NO
Quantity 10 mg
Synonym ICA-17043
Solubility Information DMSO : 50 mg/mL (154.64 mM; Need ultrasonic)
SMILES O=C(C(C1=CC=CC=C1)(C2=CC=C(C=C2)F)C3=CC=C(F)C=C3)N
Molecular Weight (g/mol) 323.34
Formula Weight 323.34
Percent Purity 98.08%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.