missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP2 Antibody (OTI4B3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74060V
This item is not returnable.
View return policy
Description
SENP2 Monoclonal specifically detects SENP2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SENP2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SENP2 | |
| Recombinant protein fragment corresponding to amino acids SENP2(NP_067640) produced in E.coli. | |
| 0.1 mL | |
| 59343.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI4B3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction