missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP2 Antibody (OTI4B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45759
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
SENP2 Monoclonal specifically detects SENP2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| SENP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunofluorescence 1:100 | |
| AXAM2Axam2, DKFZp762A2316, EC 3.4.22, EC 3.4.22.68, KIAA1331sentrin-specific protease 2, Sentrin/SUMO-specific protease SENP2, Smt3ip2, SMT3IP2EC 3.4.22.-, SMT3-specific isopeptidase 2, SUMO1/sentrin/SMT3 specific peptidase 2, SUMO1/sentrin/SMT3 specific protease 2 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 59343 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| OTI4B3 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| SENP2 | |
| Human recombinant protein fragment corresponding to amino acids SENP2(NP_067640) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction