missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP2 Monoclonal Antibody (OTI3D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800129
This item is not returnable.
View return policy
Description
SUMO is a small ubiquitin-like protein that can be covalently conjugated to other proteins. SENP2 is one of a group of enzymes that process newly synthesized SUMO1, SUMO2, and SUMO3 into the conjugatable mature forms and catalyze the deconjugation of these same SUMO proteins from their targeted substrates. SENP2 may also down-regulate CTNNB1 levels and thereby modulate the Wnt pathway.Specifications
| SENP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HC62 | |
| SENP2 | |
| Human recombit protein fragment corresponding to amino acids 139-523 of human SENP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3D12 | |
| Unconjugated | |
| SENP2 | |
| AXAM2, SMT3IP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 59343 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction