missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SENP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584450
This item is not returnable.
View return policy
Description
Immunogen sequence: LREEHVTCVQ SILDEFLQTY GSLIPLSTDE VVEKLEDIFQ QEFSTPSRKG LVLQLIQSYQ RMPGNAMVRG FRVAYKRHVL TMDDLGTL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates.
Specifications
| SENP3 | |
| Polyclonal | |
| Unconjugated | |
| Senp3 | |
| AA408656; DKFZP586K0919; DKFZp762A152; SENP3; sentrin/SUMO-specific protease 3; sentrin/SUMO-specific protease SENP3; Sentrin-specific protease 3; Smt3ip; Smt3ip1; Smt3-specific isopeptidase 1; SSP3; SUMO specific peptidase 3; SUMO/sentrin specific peptidase 3; SUMO/sentrin specific protease 3; SUMO1/sentrin/SMT3 specific peptidase 3; SUMO1/sentrin/SMT3 specific protease 3; SUMO-1-specific protease 3; SUSP3; Ulp1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4L4 | |
| Senp3 | |
| Recombinant protein corresponding to Human SENP3. Recombinant protein control fragment (Product #RP-101397). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction