missing translation for 'onlineSavingsMsg'
Learn More

SENP7 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18445473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18445473 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18445473 Supplier Novus Biologicals Supplier No. NBP150027AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SENP7 Polyclonal antibody specifically detects SENP7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP7
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.22.-, KIAA1707SUMO1/sentrin specific protease 7, MGC157730, Sentrin/SUMO-specific protease SENP7, sentrin-specific protease 7, SSP2, SUMO1/sentrin specific peptidase 7, SUMO-1-specific protease 2, SUSP2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 650 and 700 of human SUMO1/Sentrin Specific Peptidase 7 using the numbering given in entry EAW79800.1 (GeneID 57337).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57337
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.