missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP8 Monoclonal Antibody (OTI10C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805442
This item is not returnable.
View return policy
Description
Principal Names: Cysteine protease FKSG8; DEN1; FKSG8; HsT17512; NEDP1; PRSC2; SUMO/sentrin specific protease family member 8; Official Gene Symbol: SENP8 Gene ID: 123228 (Human) Gene Map Locus: 15q23 (Human) SENP8, is a novel cysteine protease which belongs to the ULP family of deubiquitinases. ULP family members consist of a C-terminal core catalytic domain and N-terminal extensions determining substrate specificity. SENP8, a 24KDa protein, plays a major role in regulating NEDD8 pathway. It processes preNEDD8 to its mature form and deconjugates NEDD8 from substrates such as Cullins and p53. SENP8 may play a role in preventing accumulation of hyper-neddylated cullin and maintaining the level of mono-neddylated cullin that is thought to increase the efficiency of ubiquitination by SCF ubiquitin ligases.Specifications
| SENP8 | |
| Monoclonal | |
| Unconjugated | |
| SENP8 | |
| DEN1, NEDP1, PRSC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 123228 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI10C11 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96LD8 | |
| SENP8 | |
| Full length human recombit protein of human SENP8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction