Learn More
LNI Swissgas SENS Hydrogen C4
Product Code. 17851969 Shop All LNI Swissgas Products
Click to view available options
Type:
Hydrogen Sensor Cable
Unit Size:
Each
Product Code. 17851969
Brand: LNI Swissgas 6940.00.160
Cable for hydrogen sensor.
  • 2 m length cable for hydrogen sensor with 4 pins.
Specifications
Type Hydrogen Sensor Cable