missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sensititre™ Extended Spectrum Beta-lactamase Plate
Click to view available options
Quantity:
10/Pk.
Description
Obtain accurate susceptibility results for Enterobacteriacea, as well as ESBL Confirmation, with the Thermo Scientific™ Sensititre™ Extended Spectrum Beta-lactamase Plate. Extensive dilution ranges of confirmatory antimicrobics ensure full compliance with NCCLS recommendations, and a comprehensive selection of antimicrobials aid in selecting the most appropriate therapeutic treatment.
Obtain accurate susceptibility results for Enterobacteriacea, as well as ESBL Confirmation, with the Sensititre Extended Spectrum Beta-lactamase Plate.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
- Manual or automated read options provide flexible alternatives for fully automated or back-up test procedures
- Long shelf life and room-temperature storage simplify inventory control and reduce expiry costs
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| 0.5 McFarland Standard | H2O (Part No. T3339) |
| Antimicrobial Agents | Various |
| AST Type | MIC |
| Broth Type | MHB (Part No. T3462) |
| Description | Sensititre Extended Spectrum Beta-lactamase Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Non-fastidious, Gram-negative Organisms |
| Test Type | Minimum Inhibitory Concentration (MIC), Breakpoint |
| Incubation Conditions | 35°C O2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction