Learn More
Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate
Description
Perform accurate AST of non-fastidious Gram-negative isolates with this convenient plate
Confidently perform susceptibility testing of non-fastidious Gram-negative isolates with Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate.
Confidently perform susceptibility testing of non-fastidious Gram-negative isolates with Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate.
- Automated or manual read options are available for ultimate flexibility
- Extended shelf life and room-temperature storage simplify inventory control
- Individual packaging allows the laboratory to test one plate at a time with no waste
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| 0.5 McFarland Standard | H2O (Part No. T3339) |
| Antimicrobial Agents | Various |
| AST Type | MIC |
| Broth Type | MHB (Part No. T3462) |
| Concentration | Multiple |
| Description | Sensititre Gram Negative MIC Plate, with Doripenem and Tigecycline |
| Antibiotic Code | Various |
| Format | Multi-antibiotic Microtitre Panels |
| Product Type | AST Plate |
| Target Organism Class | Non-fastidious, Gram-negative Organisms |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.