Learn More
WTW™ SenTix™ Mic PLUS Special pH Electrode
Click to view available options
Connector Types:
Plug Head
Unit Size:
Each
Description
- pH micro electrode, 5 mm membrane
- Specialists for all cases, pH electrodes for special applications can determine the pH value of surfaces, suspended solids, suspensions and much more
- Wide selection of special measurements
- Adaptation to the sample consistency by special glass membranes
- Optimal measuring accuracy by adapted diaphragm technique
- Micro special electrode that can solve nearly any pH measuring problem
Specifications
Specifications
| Color | Black |
| Description | PermaSleeve Wire Marking Sleeves |
| Connector Types | Plug Head |
| Junction | Ceramic |
| Material | Glass Shaft, Ceramic Diaphragm |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | SenTix™ PLUS Special pH Electrode |
| For Use With (Equipment) | Vario pH meter |
| Includes | DIN, BNC electrode plug |
| Show More |