missing translation for 'onlineSavingsMsg'
Learn More

Sepantronium bromide, MedChemExpress

Product Code. 16453648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453648 10 mg 10mg
16490037 50 mg 50mg
16416387 5 mg 5mg
16414736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453648 Supplier Medchem Express Supplier No. HY10194

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sepantronium bromide (YM-155) is a survivin inhibitor with an IC50 of 0.54 nM.

Sepantronium bromide (YM-155) is a survivin inhibitor with an IC50 of 0.54 nM.

Specifications

Chemical Name or Material Sepantronium bromide
CAS 781661-94-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19BrN4O3
Quantity 10 mg
Synonym YM-155
Solubility Information H2O : 100 mg/mL (225.59 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (112.79 mM; Need ultrasonic)
SMILES O=C1C2=C(C(C3=CC=CC=C31)=O)[N+](CC4=NC=CN=C4)=C(C)N2CCOC.[Br-]
Molecular Weight (g/mol) 443.29
Formula Weight 443.29
Percent Purity 96.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.