missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Separase Antibody (XJ11-1B12), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-439V
This item is not returnable.
View return policy
Description
Separase Monoclonal specifically detects Separase in Human samples. It is validated for Western Blot.
Specifications
| Separase | |
| Monoclonal | |
| DyLight 405 | |
| Q14674 | |
| ESPL1 | |
| Maltose-Binding Protein fusion of a C-terminal fragment of human Separase (residues 1866-1996). [UniProt# Q14674] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, Mitotic Regulators, Phospho Specific | |
| 9700 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot | |
| XJ11-1B12 | |
| Western Blot | |
| Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction