missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Separase Antibody (XJ11-1B12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-439DL594
This item is not returnable.
View return policy
Description
Separase Monoclonal antibody specifically detects Separase in Human samples. It is validated for Western Blot
Specifications
| Separase | |
| Monoclonal | |
| DyLight 594 | |
| Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase | |
| Maltose-Binding Protein fusion of a C-terminal fragment of human Separase (residues 1866-1996). [UniProt# Q14674] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, Mitotic Regulators | |
| 9700 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot | |
| XJ11-1B12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction