Learn More
Heidolph™ Separatory Funnel Attachment
Product Code. 10100981 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10100981 Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10100981 Supplier Heidolph™ Supplier No. 5497800000
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Accessory for Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
Suitable for 4 separatory funnels (50 mL, conical) or 4 separatory funnels (100 mL, conical) up to a total length from stopper to (including) stopcock of 15.5 cm
TRUSTED_SUSTAINABILITY
Specifications
Holds 4 separatory funnels (50 mL, conical) or 4 separatory funnels (100 mL, conical) up to a total length from stopper to (including) stopcock of 15.5 cm
Product Type Separatory Funnel Attachment
For Use With (Equipment) Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020