Learn More
Invitrogen™ SEPT11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114098
Description
Immunogen sequence: MAVAVGRPSN EELRNLSLSG HVGFDSLPDQ LVNKSTSQGF CFNILCVGET For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
Specifications
| SEPT11 | |
| Polyclonal | |
| Unconjugated | |
| SEPT11 | |
| 6230410I01Rik; AW548875; D5Ertd606e; SEPT11; Sept6; septin 11; septin 6; Septin11; septin-11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55752 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NVA2 | |
| SEPT11 | |
| Synthetic peptide directed towards the N terminal region of human SEPT11(septin 11) (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.