missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEPT12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598593
This item is not returnable.
View return policy
Description
SEPT12 Polyclonal Antibody for ELISA, ICC/IF, Western Blot
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).
Specifications
| SEPT12 | |
| Polyclonal | |
| Unconjugated | |
| SEPT12 | |
| 1700028G04Rik; 4933413B09Rik; SEPT12; septin 12; SEPTIN12; septin-12; SPGF10; testicular tissue protein Li 168 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 124404, 71089 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IYM1 | |
| SEPT12 | |
| Recombinant Human Septin-12 protein (1-358AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction