missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-1 Antibody (OTI1E1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74072AF700
This item is not returnable.
View return policy
Description
Septin-1 Monoclonal specifically detects Septin-1 in Human samples. It is validated for Western Blot.
Specifications
| Septin-1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SEPT1 | |
| Full length recombinant protein of human Septin-1 (NP_443070) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot | |
| OTI1E1 | |
| Western Blot | |
| DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 1731.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction