missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMCN1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06583APC
This item is not returnable.
View return policy
Description
HMCN1 Polyclonal antibody specifically detects HMCN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HMCN1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ARMD1, FBLN6, FIBL6, FIBL-6, hemicentin 1, hemicentin-1 | |
| Produced in rabbits immunized with E. coli-derived Human HMCN1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83872 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction