missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20298
This item is not returnable.
View return policy
Description
Septin-10 Polyclonal specifically detects Septin-10 in Human samples. It is validated for Western Blot.
Specifications
| Septin-10 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FLJ11619, septin 10, septin-10 | |
| Rabbit | |
| 53 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 151011 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000 | |
| Q9P0V9 | |
| 43353 | |
| Recombinant protein encompassing a sequence within the center region of human SEPT10. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (87%), Rat (86%), Bovine (88%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction