missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31717
This item is not returnable.
View return policy
Description
Septin-11 Polyclonal specifically detects Septin-11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Septin-11 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| 43354 | |
| Recombinant protein encompassing a sequence within the center region of human Septin 11. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.37mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FLJ10849, septin 11, septin-11 | |
| Rabbit | |
| 49 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55752 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction