missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
€ 271.00 - € 533.00
Specifications
| Antigen | Septin-2 |
|---|---|
| Dilution | Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18486731
|
Novus Biologicals
NBP1-85212-25ul |
25 ul |
€ 271.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776763
|
Novus Biologicals
NBP1-85212 |
0.1 mL |
€ 533.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-2 Polyclonal specifically detects Septin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Septin-2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q15019 | |
| 4735 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WGVVEVENPEHNDFLKLRTMLITHMQDLQEVTQDLHYENFRSERLKRGGRKVENEDMNKDQILLEKEAELRRMQEMIARMQAQMQMQMQGGDGDGGALGH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 41 kDa |
| Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DIFF6NEDD-5, hNedd5, KIAA0158Neural precursor cell expressed developmentally down-regulated protein 5, NEDD5Pnutl3, neural precursor cell expressed, developmentally down-regulated 5, septin 2, septin-2 | |
| 43345 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Septin-2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title