missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin 3 Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811195S
This item is not returnable.
View return policy
Description
Septin 3 Monoclonal antibody specifically detects Septin 3 in Bacteria samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| Septin 3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| Bacteria | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C8 | |
| Unconjugated | |
| 3110018K01Rik; 3-Sep; AV154067; B530002E20Rik; bK250D10.3; CTA-250D10.3; G-septin; G-septin alpha; MGC133218; neuronal-specific septin 3; neuronal-specific septin-3; OTTHUMP00000028742; P40; Sep3; SEPT3; septin 3; Septin3 | |
| Human recombinant protein fragment corresponding to amino acids 1-1166 of human CAS9 produced in E.coli. | |
| 30 μL | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction