missing translation for 'onlineSavingsMsg'
Learn More

Septin 3 Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene

Product Code. p-200016814
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17564984 30 μL 30µL
17544984 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17564984 Supplier Origene Technologies Supplier No. TA811196S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Septin 3 Monoclonal antibody specifically detects Septin 3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Septin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SEPT3
Gene Accession No. Q9UH03, Q9Z1S5
Gene Alias 3110018K01Rik; 3-Sep; AV154067; B530002E20Rik; bK250D10.3; CTA-250D10.3; G-septin; G-septin alpha; MGC133218; neuronal-specific septin 3; neuronal-specific septin-3; OTTHUMP00000028742; P40; Sep3; SEPT3; septin 3; Septin3
Host Species Mouse
Immunogen Full length human recombinant protein of human SEPT3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24050, 55964
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.