missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555244
This item is not returnable.
View return policy
Description
Immunogen sequence: KPMEDEDRYT TCWPKTWLAK DCPALRIISV EYDTSLSDWR ARCPMERKSI AFRSNELLRK LRAAGVGDRP VVWISHSMGG LLVKKMLLEA STKPEMS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The function of this protein remains unknown.
Specifications
| SERAC1 | |
| Polyclonal | |
| Unconjugated | |
| Serac1 | |
| 4930511N22Rik; D17Ertd141e; hypothetical protein LOC519860; protein SERAC1; Protein SERAC1-like protein; RGD1565064; Serac1; serine active site containing 1; serine active site-containing protein 1; srac1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84947 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JX3 | |
| Serac1 | |
| Recombinant protein corresponding to Human SERAC1. Recombinant protein control fragment (Product #RP-94734). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction