missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SERBP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15515002
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515002 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515002 Supplier Invitrogen™ Supplier No. PA541741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: PNEGADGQWK KGFVLHKSKS EEAHAEDSVM DHHFRKPAND ITSQLEINFG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SERBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SERBP1
Gene Accession No. Q8NC51
Gene Alias 1200009K13Rik; 9330147J08Rik; AL022786; CGI-55; CHD3IP; chromodomain helicase DNA binding protein 3 interacting protein; HABP4L; hypothetical RNA binding protein RDA288; MGC128417 protein; PAI RBP 1; PAI-1 mRNA binding protein; PAI-1 mRNA-binding protein; PAI1 RNA-binding protein 1; Pairbp1; PAI-RBP1; plasminogen activator inhibitor 1 RNA binding protein; plasminogen activator inhibitor 1 RNA-binding protein; Rda288; RNA binding protein RDA288; SERBP1; Serpine1 mRNA binding protein 1; SERPINE1 mRNA-binding protein 1
Gene Symbols SERBP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SERBP1 (aa 311-360).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.