Learn More
SERCA2 ATPase Antibody (2A7-A1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
Brand: Novus Biologicals NB300-581-10uL
This item is not returnable.
View return policy
Description
SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SERCA2 ATPase | |
| Monoclonal | |
| Unconjugated | |
| O46674 | |
| ATP2A2 | |
| Purified canine cardiac sarcoplasmic reticulum vesicles. | |
| 10 μL | |
| Primary | |
| Detects sarcoplasmic or endoplasmic reticulum calcium (SERCA) 2 ATPase. NB300-581 recognizes the SERCA2a and SERCA2b isoforms identically. However, the antibody does not recognize the skeletal muscle (SERCA1) or housekeeping isoforms (SERCA3). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| 2A7-A1 | |
| Western Blot 1:1000, Flow Cytometry 1 μg/ 106 cells, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:100 | |
| ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2 | |
| Mouse | |
| Unpurified | |
| RUO | |
| 488 | |
| Human | |
| Ascites |