Learn More
Invitrogen™ SERF1A Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570803
Product Code. 15839213
  • € 516.00 / 100µL
Description
This target displays homology in the following species: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
This gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The duplication region includes both a telomeric and a centromeric copy of this gene. Deletions of this gene, the telomeric copy, often accompany deletions of the neighboring SMN1 gene in spinal muscular atrophy (SMA) patients, and so it is thought that this gene may be a modifier of the SMA phenotype. The function of this protein is not known; however, it bears low-level homology with the RNA-binding domain of matrin-cyclophilin, a protein which colocalizes with small nuclear ribonucleoproteins (snRNPs) and the SMN1 gene product. Alternatively spliced transcripts have been documented but it is unclear whether alternative splicing occurs for both the centromeric and telomeric copies of the gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SERF1A
Polyclonal
Unconjugated
SERF1A
4F5; FAM2A; FAM2B; h4F5; protein 4F5; SERF1; SERF1A; SERF1B; SMA modifier 1; small EDRK-rich factor 1; small EDRK-rich factor 1A; small EDRK-rich factor 1A (telomeric); small EDRK-rich factor 1B (centromeric); SMAM1; spinal muscular atrophy-related gene H4F5; Unknown (protein for MGC:133508)
Rabbit
Affinity Chromatography
RUO
8293
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O75920
SERF1A
synthetic peptide directed towards the N-terminal region of human SERF1A.