missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERF2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SERF2.
SERF2 (small EDRK-rich factor 2) is the human ortholog of the C/ elegans MOAG-4 gene. Serf2 (small EDRK-rich factor 2) is the human ortholog of the C. elegans MOAG-4 gene. MOAG-4 was shown to be a positive regulator of protein aggregates. Inactivation of MOAG-4 leads to a rescue from proteotoxcity, and could play a role in human diseases such as Alzheimer's disease and Parkinson's disease. In addition, SERF2 can induce huntingtin aggregation in SK-N-SH neuroblastoma cells.
Specifications
Specifications
| Antigen | SERF2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SERF2 |
| Gene Accession No. | P84101, P84102 |
| Gene Alias | 4F5REL; 4F5rel (4F5 related); C80083; FAM2C; Gastric cancer-related protein VRG107; H4F5REL; HsT17089; m4F5rel; Msmac1l; Protein 4F5-related; SERF2; Small EDRK-rich factor 2 |
| Gene Symbols | SERF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?