missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Serglycin Polyclonal Antibody
GREENER_CHOICE

Product Code. 15739410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15739410 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15739410 Supplier Invitrogen™ Supplier No. PA551452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MQKLLKCSRL VLALALILVL ESSVQGYPTR RARYQWVRCN PDSNSANCLE EKGPMFELLP GESNKIPRLR TDLFPKTRIQ DLNRIFPLSE DYSGSGFGSG SGSGSGSGSG FLTEMEQDYQ LVDE Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 55%.

This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. Two transcript variants, only one of them protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Serglycin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0,25 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRGN
Gene Accession No. P10124
Gene Alias gp600; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; hematopoietic proteoglycan core protein; Mastocytoma proteoglycan core protein; p.PG; Platelet proteoglycan core protein; PPG; Prg; Prg1; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; proteoglycan, secretory granule; secretory granule proteoglycan core peptide; secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan; Sgc; Srgn
Gene Symbols SRGN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Serglycin. Recombinant protein control fragment (Product #RP-93682).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.