Learn More
3M™ Series G2000 Ventilated Hard Hat with Leather Sweatband
Description
- A hard hat with excellent ventilation a good field of vision and one of few lightweight helmets available
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it
- Extended edge on the back of the helmet provides extra protection for the sensitive neck area
- Flat surface for printing a logotype, brand name, etc
- Easy to replace sweatband for comfort and hygiene
- Ventilated headband for improved comfort
Specifications
Specifications
| Certifications/Compliance | EN 397 |
| Color | White |
| Material | UV-stabilized ABS |
| Product Type | Helmet |
| Size | 54 to 62 cm |
| Suspension Type | Pinlock |
| Weight (Metric) | 340 g |
| Ventilated | Ventilated |