missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERINC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97673-100ul
This item is not returnable.
View return policy
Description
SERINC3 Polyclonal antibody specifically detects SERINC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SERINC3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| AIGP1, DIFF33tumor differentially expressed 1, placental transmembrane protein (mouse testicular tumor differentiallyexpressed), SBBI99, serine incorporator 3, TDE, TDE1Tumor differentially expressed protein 1, TMS-1, transmembrane protein SBBI99, tumour differentially expressed 1 | |
| Produced in rabbits immunized with E. coli-derived Human SERINC3 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10955 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction