missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine Dehydratase Antibody (OTI3D3), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74074C
This item is not returnable.
View return policy
Description
Serine Dehydratase Monoclonal specifically detects Serine Dehydratase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Serine Dehydratase | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SDS | |
| Full length recombinant protein of human Serine Dehydratase (NP_006834) produced in HEK293T cells. | |
| 0.1 mL | |
| 10993.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3D3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 4.3.1.17, EC 4.3.1.19, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDHL-serine dehydratase, serine dehydratase, TDH | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction