Learn More
Serine Dehydratase Antibody (OTI3D3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74074PCP
Product Code. 18172956
  • € 785.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Serine Dehydratase Monoclonal specifically detects Serine Dehydratase in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Serine Dehydratase
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
SDS
Full length recombinant protein of human Serine Dehydratase (NP_006834) produced in HEK293T cells.
0.1 mL
10993.0
Store at 4°C in the dark.
Flow Cytometry
OTI3D3
Flow Cytometry
EC 4.3.1.17, EC 4.3.1.19, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDHL-serine dehydratase, serine dehydratase, TDH