Learn More
Serine protease 23 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-06049MFV610
Product Code. 18474439
  • € 486.00 / 0.10mL
Description
Serine protease 23 Polyclonal antibody specifically detects Serine protease 23 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry (Paraffin)
Produced in rabbits immunized with E. coli-derived Human Serine protease 23 fragment.
Cell Biology, Cytoskeleton Markers, Signal Transduction
Store at 4C in the dark.
mFluor Violet 610 SE