missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine racemase Antibody (OTI2E3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74083DL594
This item is not returnable.
View return policy
Description
Serine racemase Monoclonal antibody specifically detects Serine racemase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Serine racemase | |
| Monoclonal | |
| DyLight 594 | |
| D-serine ammonia-lyase, D-serine dehydratase, EC 4.3.1.17, EC 4.3.1.18, EC 5.1.1.18, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, serine racemase | |
| Full length recombinant protein of human Serine racemase (NP_068766) produced in HEK293T cells. | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 63826 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2E3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction