missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin Antibody (YC5/45), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65037B
This item is not returnable.
View return policy
Description
Serotonin Monoclonal specifically detects Serotonin in All Species samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
Specifications
| Serotonin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein G purified | |
| Primary | |
| All species | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| YC5/45 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount | |
| 5-HT, 5-hydroxytryptamne | |
| Serotonin conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| NB100-65037 recognizes serotonin and does not cross react with 5-hydroxyindolacetic acid, GABA, noradrenaline, 5-hydroxytryptophan, carnosine or melatonin. Serotonin (also known as 5-hydroxytryptamine or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotonin is widely distributed in the central and peripheral nervous system, and also in some non-neural tissues such as platelets. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction